Proteintech
Manufacturer Code:116231AP150UL
Catalog # 50-172-7319
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PRKD2 Fusion Protein Ag2192 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HSPC187 PKD2 PRKD2 protein kinase D2; nPKC-D2; Serine/threonine-protein kinase D2
Gene Aliases: AI325941; HSPC187; nPKC-D2; PKD2; PRKD2; RGD1308054
UniProt ID: (Human) Q9BZL6, (Rat) Q5XIS9, (Mouse) Q8BZ03
Entrez Gene ID: (Human) 25865, (Rat) 292658, (Mouse) 101540
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.