Proteintech
Manufacturer Code:121481AP150UL
Catalog # 50-172-7748
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PRKCSH Fusion Protein Ag2796 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 80K H protein AGE R2 G19P1 Glucosidase 2 subunit beta Glucosidase II subunit beta PCLD PKCSH PLD1 PRKCSH; 80K-H protein; advanced glycation end-product receptor 2; AGE-binding receptor 2; alpha glucosidase II, beta-subunit; carbohydrate processing enzyme of the endoplasmic reticulum; Glucosidase 2 subunit beta; glucosidase II beta-subunit; Glucosidase II subunit beta; hepatocystin; PKCSH; Protein kinase C substrate 60.1 kDa protein heavy chain; protein kinase C substrate, 80 Kda protein
Gene Aliases: 80K-H; AGE-R2; G19P1; GIIB; PCLD; PKCSH; PLD1; PRKCSH; VASAP-60
UniProt ID: (Human) P14314, (Mouse) O08795
Entrez Gene ID: (Human) 5589, (Mouse) 19089, (Rat) 300445
Molecular Function: enzyme modulator transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.