OriGene
Manufacturer Code:CF502415
Catalog # 50-166-1321
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4G8 |
Immunogen | Full length human recombit protein of human PRKCE produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: nPKC-epsilon; PKCE nPKC-epsilon; Protein kinase C epsilon type; protein kinase C, epsilon
Gene Aliases: nPKC-epsilon; PKCE; PRKCE
UniProt ID: (Human) Q02156
Entrez Gene ID: (Human) 5581
Molecular Function:
annexin
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
kinase
non-receptor serine/threonine protein kinase
protein kinase
transfer/carrier protein
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.