Invitrogen
Manufacturer Code:MA120070
Catalog # PIMA120070
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | RS30 |
Immunogen | Purified regulatory subunit of cAMP-dependent protein kinase type II from bovine brain. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cAMP-dependent protein kinase regulatory subunit RII alpha; cAMP-dependent protein kinase type II-alpha regulatory subunit; cAMP-dependent protein kinase, regulatory subunit alpha 2; MGC3606 PKR2 PRKAR2; protein kinase A, RII-alpha subunit; protein kinase, cAMP-dependent, regulatory subunit type II alpha; protein kinase, cAMP-dependent, regulatory, type 2, alpha; unnamed protein product
Gene Aliases: PKR2; PRKAR2; PRKAR2A
UniProt ID: (Bovine) P00515, (Human) P13861
Entrez Gene ID: (Pig) 397493, (Bovine) 100139910, (Human) 5576
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.