Proteintech
Manufacturer Code:107731AP150UL
Catalog # 50-172-6585
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PRIM1 Fusion Protein Ag1124 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA primase 1; DNA primase 49 kDa subunit; DNA primase 49 kDa subunit DNA primase small subunit p49 PRIM1; DNA primase small subunit; DNA primase subunit 48; p49; primase p49 subunit; primase polypeptide 1, 49kDa; primase, DNA, polypeptide 1 (49kDa)
Gene Aliases: AI324982; p49; PRIM1
UniProt ID: (Human) P49642, (Mouse) P20664
Entrez Gene ID: (Human) 5557, (Mouse) 19075, (Rat) 246327
Molecular Function: RNA binding protein nucleic acid binding primase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.