Invitrogen
Manufacturer Code:PA572728
Catalog # PIPA572728
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PRICKLE1 antibody for a 19 amino acid synthetic peptide near the carboxy terminus of human PRICKLE1. The immunogen is within the last 50 amino acids of PRICKLE1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: prickle homolog 1; prickle like 1; prickle-like 1; Prickle-like protein 1; REST (RE-1 silencing transcription factor)/NRSF (neuron-restrictive silencer factor)-interacting LIM domain protein; REST/NRSF interacting LIM domain protein; REST/NRSF-interacting LIM domain protein 1
Gene Aliases: 1110058P22Rik; AW215793; b2b019Clo; EPM1B; mpk1; Prickle; PRICKLE1; RILP
UniProt ID: (Human) Q96MT3, (Mouse) Q3U5C7
Entrez Gene ID: (Human) 144165, (Rat) 315259, (Mouse) 106042
Molecular Function: actin family cytoskeletal protein cytoskeletal protein structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.