Novus Biologicals
Manufacturer Code:NBP183967
Catalog # NBP183967
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:GASGYNHDETQWYEDSLECLSDLKPEQSVRDSMDSLALSNITGASVDGENKPRPSLYSLQNFEEMETEDCEKMSNMGTLNSSMLHRSAESLKSLSSELCPEKILPEEKPVHLPVLRRSKSQSRPQQVKFSDDVIDNGNYDIEI |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EPM1BMGC138902 FLJ31627 FLJ31937 prickle homolog 1 (Drosophila) prickle-like 1 prickle-like 1 (Drosophila) prickle-like protein 1 REST (RE-1 silencing transcription factor)/NRSF (neuron-restrictive silencerfactor)-interacting LIM domain protein REST/NRSF-interacting LIM domain protein 1 RILPMGC138903; prickle homolog 1; prickle-like 1; Prickle-like protein 1; REST (RE-1 silencing transcription factor)/NRSF (neuron-restrictive silencer factor)-interacting LIM domain protein; REST/NRSF interacting LIM domain protein; REST/NRSF-interacting LIM domain protein 1
Gene Aliases: EPM1B; PRICKLE1; RILP
UniProt ID: (Human) Q96MT3
Entrez Gene ID: (Human) 144165
Molecular Function: actin family cytoskeletal protein cytoskeletal protein structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.