Invitrogen
Manufacturer Code:PA571241
Catalog # PIPA571241
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of human PREX2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DEP domain containing 2; DEP domain-containing protein 2; DEPDC2; P-Rex2; Phosphatidylinositol 3,4,5-trisphosphate-dependent Rac exchanger 2 protein; protein phosphatase 1, regulatory subunit 129; PtdIns(3,4,5)-dependent Rac exchanger 2
Gene Aliases: 6230420N16Rik; AI316880; AI553603; C030045D06Rik; D430013K02; DEP.2; DEPDC2; P-REX2; PPP1R129; PREX2; si:ch211-278g15.1
UniProt ID: (Human) Q70Z35, (Mouse) Q3UQI2
Entrez Gene ID: (Bovine) 520704, (Rabbit) 100346695, (Horse) 100063148, (Dog) 486984, (Pig) 100520557, (Human) 80243, (Mouse) 109294, (Rat) 312912, (Zebrafish) 557288
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.