Novus Biologicals
Manufacturer Code:NBP288090
Catalog # NB035712
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human PREX2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DEP domain-containing protein 2; DEP domain-containing protein 2 DEP.2 DEPDC2 phosphatidylinositol 345-trisphosphate-dependent Rac exchanger 2 protein phosphatidylinositol-345-trisphosphate-dependent Rac exchange factor 2 P-REX26230420N16Rik PtdIns(345)-dependent Rac exchanger 2; P-Rex2; Phosphatidylinositol 3,4,5-trisphosphate-dependent Rac exchanger 2 protein; protein phosphatase 1, regulatory subunit 129; PtdIns(3,4,5)-dependent Rac exchanger 2
Gene Aliases: DEP.2; DEPDC2; P-REX2; PPP1R129; PREX2
UniProt ID: (Human) Q70Z35
Entrez Gene ID: (Human) 80243
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.