Proteintech
Manufacturer Code:115361AP150UL
Catalog # 50-172-7244
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PREP Fusion Protein Ag2101 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dJ355L5.1 (prolyl endopeptidase); PE; PE PEP Post proline cleaving enzyme PREP prolyl endopeptidase; PE {ECO:0000250|UniProtKB:P23687}; Post-proline cleaving enzyme; post-proline cleaving enzyme {ECO:0000250|UniProtKB:P23687}; prep {ECO:0000312|RGD:620841}; Prolyl endopeptidase; prolyl endopeptidase {ECO:0000250|UniProtKB:P23687}; prolyl oligopeptidase; rPop
Gene Aliases: AI047692; AI450383; D10Wsu136e; PE; PEP; Pop; PREP; rPop
UniProt ID: (Human) P48147, (Mouse) Q9QUR6, (Rat) O70196
Entrez Gene ID: (Human) 5550, (Mouse) 19072, (Rat) 83471
Molecular Function: hydrolase protease serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.