Invitrogen
Manufacturer Code:PA5112171
Catalog # PIPA5112171
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived Human PRELID1 fragment |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 25 kDa protein of relevant evolutionary and lymphoid interest; 25 kDa protein of relevant evolutionary and lymphoid interest 2610524G07Rik CGI-106 Preli PRELI domain containing 1 PRELI domain-containing protein 1 mitochondrial PRELID1 protein of relevant evolutionary and lymphoid interest PX19 PX19 homolog Px19-like protein RGD1308082 SBBI12; PRELI domain-containing protein 1, mitochondrial; Px19-like protein
Gene Aliases: CGI-106; PRELI; PRELID1; PX19; SBBI12
UniProt ID: (Human) Q9Y255
Entrez Gene ID: (Human) 27166
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.