Proteintech
Manufacturer Code:108771AP150UL
Catalog # 50-172-6677
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PRELID1 Fusion Protein Ag1037 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 25 kDa protein of relevant evolutionary and lymphoid interest; CGI 106 PRELI PRELI domain containing 1 PRELID1 PX19 Px19 like protein SBBI12; PRELI domain-containing protein 1, mitochondrial; protein of relevant evolutionary and lymphoid interest; PX19 homolog; Px19-like protein
Gene Aliases: 2610524G07Rik; CGI-106; PRELI; PRELID1; PX19; RGD1308082; SBBI12
UniProt ID: (Human) Q9Y255, (Mouse) Q8R107
Entrez Gene ID: (Human) 27166, (Rat) 290995, (Mouse) 66494
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.