Abnova
Manufacturer Code:H00025824B01
Catalog # 89-007-344
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | PRDX5 (NP_036226.1 1 a.a. ~ 214 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human PRDX5 protein.
For Research Use Only
Protein Aliases: ACR1; Alu co-repressor 1; Alu corepressor 1; Antioxidant enzyme B166; AOEB166; B166; epididymis secretory protein Li 55; Liver tissue 2D-page spot 71B; MGC117264; MGC142283; MGC142285; Peroxiredoxin V; Peroxiredoxin-5, mitochondrial; Peroxisomal antioxidant enzyme; PLP; PMP20; PRDX6; Prx-V; PRXV; SBBI10; Thioredoxin peroxidase PMP20; thioredoxin reductase; Thioredoxin-dependent peroxiredoxin 5; TPx type VI
Gene Aliases: ACR1; AOEB166; B166; HEL-S-55; PLP; PMP20; PRDX5; PRDX6; prx-V; PRXV; SBBI10
UniProt ID: (Human) P30044
Entrez Gene ID: (Human) 25824
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.