Proteintech
Manufacturer Code:550871AP150UL
Catalog # 50-173-5974
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: anti-oxidant protein 1; antioxidant protein; Antioxidant protein 1; Antioxidant protein 1 AOP 1 AOP1 HBC189 MER5 Peroxiredoxin 3 Peroxiredoxin III PRDX3 PRO1748 Protein MER5 homolog Prx III PRX3 SP 22; AOP-1; HBC189; mitochondrial thioredoxin dependent peroxide reductase; mitochondrial Trx dependent peroxide reductase; Perioredoxin-3; Peroxiredoxin III; Peroxiredoxin-3; Protein MER5; Protein MER5 homolog; PRX III; PRX-3; Prx-III; Thioredoxin-dependent peroxide reductase, mitochondrial; Thioredoxin-dependent peroxiredoxin 3
Gene Aliases: AOP-1; AOP1; AW822249; D0Tohi1; Ef2l; HBC189; MER5; PRDX3; PRO1748; prx-III; Prx3; SP-22; SP22; TDXM
UniProt ID: (Human) P30048, (Mouse) P20108, (Rat) Q9Z0V6
Entrez Gene ID: (Human) 10935, (Mouse) 11757, (Rat) 64371
Molecular Function:
oxidoreductase
peroxidase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.