Novus Biologicals
Manufacturer Code:NBP232478
Catalog # NBP232478
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: QELGIKWGSKWKKELMAGREPKPEIHPCPSCCLAFSSQKFLSQHVERNHSSQNFPGPSARKLLQPENPCPG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [histone H3]-lysine36 N-trimethyltransferase PRDM9; [histone H3]-lysine4 N-trimethyltransferase PRDM9; [histone H3]-lysine9 N-trimethyltransferase PRDM9; [histone H4]-lysine20 N-methyltransferase PRDM9; [histone H4]-N-methyl-L-lysine20 N-methyltransferase PRDM9; EC 2.1.1.43 histone-lysine N-methyltransferase PRDM9 minisatellite binding protein 3 (115kD) minisatellite binding protein 3 115kDa PFM6MEISETZ PR domain containing 9 PR domain zinc finger protein 9 PR domain-containing protein 9 PRMD9 ZNF899; Histone-lysine N-methyltransferase PRDM9; minisatellite binding protein 3 (115kD); PR domain containing 9; PR domain zinc finger protein 9; PR domain-containing protein 9; Protein-lysine N-methyltransferase PRDM9
Gene Aliases: MEISETZ; MSBP3; PFM6; PRDM9; ZNF899
UniProt ID: (Human) Q9NQV7
Entrez Gene ID: (Human) 56979
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.