Novus Biologicals
Manufacturer Code:NBP29431320UL
Catalog # NB023181
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 376-595 of human PRDM6 (NP_001129711.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PR domain containing 6; PR domain zinc finger protein 6; PR domain-containing protein 6; PR-domain zinc finger protein 6; PRDM6 PR domain containing 6 putative histone-lysine N-methyltransferase PRDM6; Putative histone-lysine N-methyltransferase PRDM6
Gene Aliases: PFM3; PRDM6; PRISM
UniProt ID: (Human) Q9NQX0
Entrez Gene ID: (Human) 93166
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.