Invitrogen
Manufacturer Code:PA540606
Catalog # PIPA540606
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human PRDM5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PR domain containing 5; PR domain containing 5 PFM2 BCS2; PR domain zinc finger protein 5; PR domain-containing protein 5; un-named hi61; unm hi61
Gene Aliases: 4432417F03Rik; 6530401I24Rik; AI197291; BCS2; E130112L17Rik; PFM2; PRDM5
UniProt ID: (Human) Q9NQX1, (Mouse) Q9CXE0
Entrez Gene ID: (Bovine) 537500, (Human) 11107, (Dog) 483850, (Horse) 100072313, (Rabbit) 100341114, (Mouse) 70779, (Rat) 689788, (Zebrafish) 432371
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.