Invitrogen
Manufacturer Code:PA530430
Catalog # PIPA530430
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 559 and 758 of Human PRDM4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PFM1 PR domain zinc finger protein 4 PR domain-containing protein 4 PR-domain zinc-finger protein PFM1; PR domain containing 4; PR domain zinc finger protein 4; PR domain-containing protein 4; PR-domain zinc-finger protein PFM1
Gene Aliases: 1700031E19Rik; 2810470D21Rik; AW552272; PFM1; PRDM4; SC-1
UniProt ID: (Human) Q9UKN5, (Mouse) Q80V63
Entrez Gene ID: (Human) 11108, (Mouse) 72843
Molecular Function: KRAB box transcription factor transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.