Invitrogen
Manufacturer Code:PA599266
Catalog # PIPA599266
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PRDM12 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Gm998 HSAN8 PFM9 PR domain 12 PR domain containing 12 PR domain zinc finger protein 12 PR domain-containing protein 12 Prdm12 PR-domain containing protein 12 PR-domain zinc finger protein 12 RP11-57C19.7; PR domain containing 12; PR domain zinc finger protein 12; PR domain-containing protein 12; PR-domain containing protein 12; PR-domain zinc finger protein 12
Gene Aliases: Gm998; HSAN8; PFM9; PRDM12
UniProt ID: (Human) Q9H4Q4, (Mouse) A2AJ77
Entrez Gene ID: (Human) 59335, (Mouse) 381359
Molecular Function:
DNA binding protein
centromere DNA-binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.