Invitrogen
Manufacturer Code:MA511143
Catalog # PIMA511143
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 48AT1224.90.46 |
Immunogen | Purified recombit GST fusion protein encoding the N-terminal part of human PRDM10. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: PFM7; PR domain containing 10; PR domain zinc finger protein 10; PR domain-containing protein 10; PR-domain family member 7; PRDM zinc finger transcription factor; Tristanin
Gene Aliases: Gm1112; KIAA1231; PFM7; PRDM10; TRIS; tristanin
UniProt ID: (Human) Q9NQV6
Entrez Gene ID: (Human) 56980, (Mouse) 382066
Molecular Function:
KRAB box transcription factor
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.