Invitrogen
Manufacturer Code:PA5104031
Catalog # PIPA5104031
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PRDM10 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Gm1112 KIAA1231 MGC131802 PFM7 PR domain 10 PR domain containing 10 PR domain zinc finger protein 10 PR domain-containing protein 10 PRDM zinc finger transcription factor PRDM10 PR-domain family member 7 RGD1560384 TRIS Tristanin; PR domain containing 10; PR domain zinc finger protein 10; PR domain-containing protein 10; PR-domain family member 7; PRDM zinc finger transcription factor; Tristanin
Gene Aliases: Gm1112; KIAA1231; PFM7; PRDM10; RGD1560384; TRIS; tristanin
UniProt ID: (Human) Q9NQV6
Entrez Gene ID: (Human) 56980, (Mouse) 382066, (Rat) 500964
Molecular Function:
KRAB box transcription factor
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.