Novus Biologicals
Manufacturer Code:NBP2855260.1ML
Catalog # NB033455
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human PRDM10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1231PRDM zinc finger transcription factor MGC131802 PFM7tristanin PR domain containing 10 PR domain zinc finger protein 10 PR domain-containing protein 10 PR-domain family member 7 TRIS Tristanin; PR domain containing 10; PR domain zinc finger protein 10; PR domain-containing protein 10; PR-domain family member 7; PRDM zinc finger transcription factor; Tristanin
Gene Aliases: KIAA1231; PFM7; PRDM10; TRIS
UniProt ID: (Human) Q9NQV6
Entrez Gene ID: (Human) 56980
Molecular Function:
KRAB box transcription factor
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.