Invitrogen
Manufacturer Code:PA553543
Catalog # PIPA553543
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PRCP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Angiotensinase C; Lysosomal carboxypeptidase C; Lysosomal Pro-X carboxypeptidase; Lysosomal Pro-X carboxypeptidase Angiotensinase C Lysosomal carboxypeptidase C Proline carboxypeptidase Prolylcarboxypeptidase PRCP PRCP PCP; PRCP; Proline carboxypeptidase; Prolylcarboxypeptidase; prolylcarboxypeptidase (angiotensinase C)
Gene Aliases: HUMPCP; PCP; PRCP
UniProt ID: (Human) P42785
Entrez Gene ID: (Human) 5547
Molecular Function:
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.