Proteintech
Manufacturer Code:159951AP150UL
Catalog # 50-173-0627
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PRCP Fusion Protein Ag8869 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Angiotensinase C; Angiotensinase C HUMPCP Lysosomal carboxypeptidase C PCP PRCP Proline carboxypeptidase Prolylcarboxypeptidase; Lysosomal carboxypeptidase C; Lysosomal Pro-X carboxypeptidase; PRCP; Proline carboxypeptidase; Prolylcarboxypeptidase; prolylcarboxypeptidase (angiotensinase C)
Gene Aliases: 2510048K03Rik; 2610104A14Rik; AI451719; HUMPCP; PCP; PRCP
UniProt ID: (Human) P42785, (Mouse) Q3TLW3
Entrez Gene ID: (Human) 5547, (Rat) 293118, (Mouse) 72461
Molecular Function:
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.