Invitrogen
Manufacturer Code:MA1846
Catalog # PIMA1846
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 16F2 |
Immunogen | Recombit PRC1 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: anaphase spindle elongation 1 homolog; PRC1; protein regulating cytokinesis 1; Protein regulator of cytokinesis 1
Gene Aliases: ASE1; D7Ertd348e; PRC1
UniProt ID: (Human) O43663, (Mouse) Q99K43
Entrez Gene ID: (Human) 9055, (Mouse) 233406, (Rat) 308761
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.