Novus Biologicals
Manufacturer Code:NBP299648AF700
Catalog # NB086036
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human PRC1. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: anaphase spindle elongation 1 homolog; anaphase spindle elongation 1 homolog ASE1 MGC1671 MGC3669 protein regulating cytokinesis 1 protein regulator of cytokinesis 1; PRC1; protein regulating cytokinesis 1; Protein regulator of cytokinesis 1
Gene Aliases: ASE1; PRC1
UniProt ID: (Human) O43663
Entrez Gene ID: (Human) 9055
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.