Novus Biologicals
Manufacturer Code:NBP273594AF488
Catalog # NB273594A48
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5E1 |
Immunogen | Recombinant protein fragment corresponding to amino acids 197-471 of human PRAK (NP_003659) produced in E.coli. |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: EC 2.7.11 MAP kinase-activated protein kinase 5 MAPK-activated protein kinase 5 MAPKAP kinase 5 MAPKAPK-5 mitogen-activated protein kinase-activated protein kinase 5 p38-regulated/activated protein kinase PRAKEC 2.7.11.1; MAP kinase-activated protein kinase 5; MAPK-activated protein kinase 5; MAPKAP kinase 5; MAPKAPK-5; p38-regulated/activated protein kinase; PRAK
Gene Aliases: MAPKAP-K5; MAPKAPK5; MK-5; MK5; PRAK
UniProt ID: (Human) Q8IW41
Entrez Gene ID: (Human) 8550
Molecular Function:
cytoskeletal protein
kinase
microtubule family cytoskeletal protein
non-motor microtubule binding protein
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.