Invitrogen
Manufacturer Code:PA570989
Catalog # PIPA570989
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of human PQLC2L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PQ loop repeat containing 2-like; PQ-loop repeat-containing protein 2-like; Putative uncharacterized protein SLC66A1L; Solute carrier family 66 member 1-like
Gene Aliases: C3orf55; PQLC2L; SLC66A1L
UniProt ID: (Human) A1A4F0
Entrez Gene ID: (Human) 152078
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.