Novus Biologicals
Manufacturer Code:NBP15992720UL
Catalog # NBP15992720
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to PQLC2(PQ loop repeat containing 2) The peptide sequence was selected from the middle region of PQLC2. Peptide sequence LLFLMGMACATPLLSAAGPVAAPREAFRGRALLSVESGSKPFTRQEVIGF. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ20320 FLJ23906 FLJ26661 FLJ26665 PQ loop repeat containing 2 PQ-loop repeat-containing protein 2; Lysosomal amino acid transporter 1 homolog; PQ-loop repeat-containing protein 2; Solute carrier family 66 member 1
Gene Aliases: PQLC2; SLC66A1
UniProt ID: (Human) Q6ZP29
Entrez Gene ID: (Human) 54896
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.