Invitrogen
Manufacturer Code:PA540376
Catalog # PIPA540376
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human PQBP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 38 kDa nuclear protein containing a WW domain; Npw38; Polyglutamine tract-binding protein 1; Polyglutamine-binding protein 1; PQBP-1; scurfy candidate 2
Gene Aliases: JM26; MRX2; MRX55; MRXS3; MRXS8; NPW38; PQBP-1; PQBP1; RENS1; Sfc2; SHS
UniProt ID: (Bovine) Q2HJC9, (Human) O60828, (Rat) Q6PCT5, (Mouse) Q91VJ5
Entrez Gene ID: (Bovine) 534894, (Pig) 100620849, (Human) 10084, (Rabbit) 100345368, (Dog) 480909, (Horse) 100062415, (Rat) 302557, (Guinea pig) 100732065, (Mouse) 54633
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.