Novus Biologicals
Manufacturer Code:NBP299606DL594
Catalog # NB085844
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human PQBP1. |
Conjugate | DyLight 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 38 kDa nuclear protein containing a WW domain; 38 kDa nuclear protein containing a WW domain MRX55 MRXS3 MRXS8 Npw38 NPW38mental retardation X-linked 55 nuclear protein containing WW domain 38 kD polyglutamine binding protein 1 Polyglutamine tract-binding protein 1 polyglutamine-binding protein 1 PQBP-1 RENS1 SHS Sutherland-Haan X-linked mental retardation syndrome; Npw38; Polyglutamine tract-binding protein 1; Polyglutamine-binding protein 1; PQBP-1
Gene Aliases: JM26; MRX2; MRX55; MRXS3; MRXS8; NPW38; PQBP1; RENS1; SHS
UniProt ID: (Human) O60828
Entrez Gene ID: (Human) 10084
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.