Abnova
Manufacturer Code:PAB28529
Catalog # 89-130-039
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human PQBP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant PQBP1.
For Research Use Only
Protein Aliases: 38 kDa nuclear protein containing a WW domain; MRX55; MRXS3; MRXS8; NPW38; Polyglutamine tract-binding protein 1; Polyglutamine-binding protein 1; PQBP-1; RENS1; SHS
Gene Aliases: JM26; MRX2; MRX55; MRXS3; MRXS8; NPW38; PQBP1; RENS1; SHS
UniProt ID: (Human) O60828
Entrez Gene ID: (Human) 10084
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.