Novus Biologicals
Manufacturer Code:NBP29427220UL
Catalog # NB023100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 28-302 of human PPT2 (NP_005146.4). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C6orf8 DKFZp564P1516 EC 3.1.2 EC 3.1.2.- G14 lysosomal thioesterase PPT2 palmitoyl-protein hydrolase 2 palmitoyl-protein thioesterase 2 PPT-2 S-thioesterase G14 VE-statin2; Lysosomal thioesterase PPT2; palmitoyl-protein hydrolase 2; PPT-2; S-thioesterase G14; VE-statin2
Gene Aliases: C6orf8; G14; PPT-2; PPT2
UniProt ID: (Human) Q0P6K0
Entrez Gene ID: (Human) 9374
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.