Invitrogen
Manufacturer Code:PA512228
Catalog # PIPA512228
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 32-63 amino acids from the N-terminal region of human PPT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ceroid-palmitoyl-palmitoyl-protein thioesterase 1; Palmitoyl-protein hydrolase 1; Palmitoyl-protein thioesterase 1; Palmitoyl-protein thioesterase 1 Palmitoyl-protein hydrolase 1; PPT-1
Gene Aliases: 9530043G02Rik; AA960502; C77813; CLN1; D4Ertd184e; INCL; PPT; PPT1
UniProt ID: (Human) P50897, (Mouse) O88531
Entrez Gene ID: (Human) 5538, (Mouse) 19063
Molecular Function: esterase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.