Abnova
Manufacturer Code:PAB2508
Catalog # 89-126-753
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human PPT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of PPT1.
For Research Use Only
Protein Aliases: ceroid-palmitoyl-palmitoyl-protein thioesterase 1; CLN1; INCL; Palmitoyl-protein hydrolase 1; Palmitoyl-protein thioesterase 1; PPT; PPT-1
Gene Aliases: CLN1; INCL; PPT; PPT1
UniProt ID: (Human) P50897
Entrez Gene ID: (Human) 5538
Molecular Function:
esterase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.