Invitrogen
Manufacturer Code:PA5103671
Catalog # PIPA5103671
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PPP3R2 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calcineurin B subunit; calcineurin B subunit calcineurin B type II calcineurin B type II (19kDa) Calcineurin BII calcineurin B-like protein calcineurin subunit B type 2 CaNB2 Cblp CnB2 CNBII PP2B beta 2 PPP3R1-like Ppp3r2 PPP3RL protein phospatase 3 regulatory subunit B alpha isoform (calcineurin B type II) protein phospatase 3 regulatory subunit B alpha isoformtype 2 protein phosphatase 2B regulatory subunit 2 protein phosphatase 3 (formerly 2B) regulatory subunit B (19kD) beta isoform (calcineurin B type II) protein phosphatase 3 (formerly 2B) regulatory subunit B 19kDa beta isoform (calcineurin B type II) protein phosphatase 3 (formerly 2B) regulatory subunit B beta isoform Protein phosphatase 3 regulatory subunit B beta isoform protein phosphatase 3 regulatory subunit B beta protein phosphatase 3 regulatory subunit B alpha isoform (calcineurin B type II) protein phosphatase 3 regulatory subunit B beta protein phosphatase 3 regulatory subunit B beta isoform ty; calcineurin B, type II; calcineurin B, type II (19kDa); Calcineurin B-like protein; Calcineurin BII; Calcineurin subunit B type 2; CBLP; CNBII; PP2B beta 2; PPP3R1-like; protein phospatase 3, regulatory subunit B, alpha isoform (calcineurin B, type II); protein phospatase 3, regulatory subunit B, alpha isoform,type 2; Protein phosphatase 2B regulatory subunit 2; protein phosphatase 3 (formerly 2B), regulatory subunit B (19kD), beta isoform (calcineurin B, type II); protein phosphatase 3 (formerly 2B), regulatory subunit B, 19kDa, beta isoform (calcineurin B, type II); protein phosphatase 3 (formerly 2B), regulatory subunit B, beta isoform; Protein phosphatase 3 regulatory subunit B beta isoform; protein phosphatase 3, regulatory subunit B, alpha isoform (calcineurin B, type II); protein phosphatase 3, regulatory subunit B, beta; protein phosphatase 3, regulatory subunit B, beta isoform
Gene Aliases: CaNB2; CBLP; CnB2; PPP3R2; PPP3RL
UniProt ID: (Human) Q96LZ3, (Mouse) Q63811, (Rat) P28470
Entrez Gene ID: (Human) 5535, (Mouse) 19059, (Rat) 29749
Molecular Function: calcium-binding protein calmodulin hydrolase intracellular calcium-sensing protein phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.