Abnova
Manufacturer Code:H00005535A01
Catalog # 89-003-058
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | PPP3R2 (NP_671709 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant PPP3R2.
For Research Use Only
Protein Aliases: calcineurin B, type II (19kDa); Calcineurin B-like protein; Calcineurin BII; Calcineurin subunit B type 2; CBLP; CNBII; PPP3R1-like; PPP3RL; Protein phosphatase 2B regulatory subunit 2; protein phosphatase 3 (formerly 2B), regulatory subunit B (19kD), beta isoform (calcineurin B, type II); protein phosphatase 3 (formerly 2B), regulatory subunit B, 19kDa, beta isoform (calcineurin B, type II); protein phosphatase 3 (formerly 2B), regulatory subunit B, beta isoform; Protein phosphatase 3 regulatory subunit B beta isoform; protein phosphatase 3, regulatory subunit B, beta
Gene Aliases: CBLP; PPP3R2; PPP3RL
UniProt ID: (Human) Q96LZ3
Entrez Gene ID: (Human) 5535
Molecular Function: calcium-binding protein calmodulin hydrolase intracellular calcium-sensing protein phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.