Invitrogen
Manufacturer Code:MA527212
Catalog # PIMA527212
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2E4 |
Immunogen | Human recombit protein fragment corresponding to amino acids 23-96 of PPP3CB produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: calcineurin A beta; calcineurin A2; Calmodulin-dependent calcineurin A subunit beta isoform; CAM-PRP catalytic subunit; CNA beta; protein phosphatase 2B, catalytic subunit, beta isoform; protein phosphatase 3 (formerly 2B), catalytic subunit, beta isoform; protein phosphatase 3, catalytic subunit, beta isozyme; protein phosphatase from PCR fragment H32; RP11-345K20.1 CALNA2 CALNB CNA2 PP2Bbeta CAM-PRP catalytic subunit calcineurin A beta calcineurin A2 calmodulin-dependent calcineurin A subunit beta isoform protein phosphatase 2B catalytic subunit beta isoform protein phosphatase 3 (formerly 2B) catalytic subunit beta isoform protein phosphatase from PCR fragment H32 serine/threonine-protein phosphatase 2B catalytic subunit beta isoform; Serine/threonine-protein phosphatase 2B catalytic subunit beta isoform
Gene Aliases: CALNA2; CALNB; CNA2; PP2Bbeta; PPP3CB
UniProt ID: (Human) P16298
Entrez Gene ID: (Human) 5532
Molecular Function: calcium-binding protein hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.