Invitrogen
Manufacturer Code:PA515581
Catalog # PIPA515581
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human PPP3CB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calcineurin A beta; calcineurin A2; Calcineurin subunit A beta; Calmodulin-dependent calcineurin A subunit beta isoform; CAM-PRP catalytic subunit; CaN Abeta; CNA beta; PP2BA beta; protein phosphatase 2B, catalytic subunit, beta isoform; protein phosphatase 3 (formerly 2B), catalytic subunit, beta isoform; protein phosphatase 3, catalytic subunit, beta isoform; protein phosphatase 3, catalytic subunit, beta isozyme; protein phosphatase from PCR fragment H32; protein phosphatase-2Bb; Serine/threonine protein phosphatase 2B catalytic subunit beta isoform Calmodulin-dependent calcineurin A subunit beta isoform CAM-PRP catalytic subunit PPP3CB CALNA2 CNA2; Serine/threonine-protein phosphatase 2B catalytic subunit beta isoform
Gene Aliases: 1110063J16Rik; CALNA2; CALNB; CNA2; Cnab; CnAbeta; PP2Bbeta; PPP3CB
UniProt ID: (Human) P16298, (Mouse) P48453, (Rat) P20651
Entrez Gene ID: (Human) 5532, (Mouse) 19056, (Rat) 24675
Molecular Function:
calcium-binding protein
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.