Abnova
Manufacturer Code:PAB2159
Catalog # 89-123-086
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human PPP3CA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of PPP3CA.
For Research Use Only
Protein Aliases: calcineurin A alpha; Calmodulin-dependent calcineurin A subunit alpha isoform; CALN; CALNA; CALNA1; CAM-PRP catalytic subunit; CCN1; CNA1; PPP2B; protein phosphatase 2B, catalytic subunit, alpha isoform; protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform; protein phosphatase 3 catalytic subunit alpha isozyme; protein phosphatase 3, catalytic subunit, alpha isozyme; Serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform
Gene Aliases: CALN; CALNA; CALNA1; CCN1; CNA; CNA1; PPP2B; PPP3CA
UniProt ID: (Human) Q08209
Entrez Gene ID: (Human) 5530
Molecular Function: calcium-binding protein hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.