Invitrogen
Manufacturer Code:PA517982
Catalog # PIPA517982
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (KRAEEFLTASQEAL) corresponding to the C-terminus amino acids of PPP2R5D |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: PP2A B subunit isoform B'-delta; PP2A B subunit isoform B56-delta; PP2A B subunit isoform PR61-delta; PP2A B subunit isoform R5-delta; PP2A, B subunit, B' delta isoform; PP2A, B subunit, B56 delta isoform; PP2A, B subunit, PR61 delta isoform; PP2A, B subunit, R5 delta isoform; PPP2R5D MGC2134 MGC8949 protein phosphatase 2 regulatory subunit B (B56) delta isoform PP2A B subunit B&prime delta isoform PP2A B subunit R5 delta isoform PP2A B subunit B56 delta isoform PP2A B subunit PR61 delta isoform Serine/threonine protein phosphatase 2A 56 kDa regulatory subunit delta isoform protein phosphatase 2 regulatory subunit B&prime delta isoform B56D delta isoform of regulatory subunit B56 protein phosphatase 2A serine/threonine-protein phosphatase 2A 56 kDa re; protein phosphatase 2, regulatory subunit B (B56), delta isoform; Serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, delta isoform; Serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit delta isoform
Gene Aliases: B56D; MRD35; PPP2R5D
UniProt ID: (Human) Q14738
Entrez Gene ID: (Human) 5528
Molecular Function:
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.