Novus Biologicals
Manufacturer Code:NB10057538
Catalog # NB10057538
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 552 and 602 of human serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit delta using the numbering given in entry NP_006236.1 (GeneID 5528). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC2134 MGC8949 PP2A B subunit isoform B56-delta PP2A B subunit isoform B'-delta PP2A B subunit isoform PR61-delta PP2A B subunit isoform R5-delta PP2A B subunit B' delta isoform PP2A B subunit B56 delta isoform PP2A B subunit PR61 delta isoform PP2A B subunit R5 delta isoform protein phosphatase 2 regulatory subunit B' delta regulatory subunit B (B56) delta isoform regulatory subunit B' delta isoform Serine/threonine protein phosphatase 2A 56 kDa regulatory subunit deltaisoform serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit delta isoform; PP2A B subunit isoform B'-delta; PP2A B subunit isoform B56-delta; PP2A B subunit isoform PR61-delta; PP2A B subunit isoform R5-delta; PP2A, B subunit, B' delta isoform; PP2A, B subunit, B56 delta isoform; PP2A, B subunit, PR61 delta isoform; PP2A, B subunit, R5 delta isoform; protein phosphatase 2, regulatory subunit B (B56), delta isoform; Serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, delta isoform; Serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit delta isoform
Gene Aliases: B56D; MRD35; PPP2R5D
UniProt ID: (Human) Q14738
Entrez Gene ID: (Human) 5528
Molecular Function:
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.