Invitrogen
Manufacturer Code:PA557740
Catalog # PIPA557740
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PPP2R5B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PP2A B subunit isoform B'-beta; PP2A B subunit isoform B56-beta; PP2A B subunit isoform PR61-beta; PP2A B subunit isoform R5-beta; PPP2R5B protein phosphatase 2 regulatory subunit B (B56) beta isoform PP2A B subunit B&prime beta isoform PP2A B subunit R5 beta isoform PP2A B subunit B56 beta isoform PP2A B subunit PR61 beta isoform serine/threonine protein phosphatase 2A 56 kDa regulatory subunit beta isoform protein phosphatase 2 regulatory subunit B&prime beta isoform PP2A B subunit R5 beta isoform beta isoform of regulatory subunit B56 protein phosphatase 2A protein phosphatase 2 regulatory subunit B (B5; protein phosphatase 2 regulatory subunit B', beta; protein phosphatase 2, regulatory subunit B (B56), beta isoform; protein phosphatase 2, regulatory subunit B', beta isoform; serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, beta isoform; Serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit beta isoform
Gene Aliases: B'beta; B56B; BC026670; LOC100909468; PPP2R5B; PR61B; rCG_47223
UniProt ID: (Human) Q15173, (Mouse) Q6PD28, (Rat) Q80W83
Entrez Gene ID: (Human) 5526, (Mouse) 225849, (Rat) 309179
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.