Invitrogen
Manufacturer Code:PA131465
Catalog # PIPA131465
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids CQRLTPQVAASGGQS from C-Terminus of Human PPP2R5B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: B56B FLJ35411 PR61B; PP2A B subunit isoform B'-beta; PP2A B subunit isoform B56-beta; PP2A B subunit isoform PR61-beta; PP2A B subunit isoform R5-beta; protein phosphatase 2 regulatory subunit B', beta; protein phosphatase 2, regulatory subunit B (B56), beta isoform; protein phosphatase 2, regulatory subunit B', beta isoform; serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, beta isoform; Serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit beta isoform
Gene Aliases: B'beta; B56B; BC026670; PPP2R5B; PR61B
UniProt ID: (Human) Q15173, (Mouse) Q6PD28
Entrez Gene ID: (Human) 5526, (Mouse) 225849
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.