Abnova
Manufacturer Code:PAB6076
Catalog # 89-132-121
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human PPP2R5B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of PPP2R5B.
For Research Use Only
Protein Aliases: B56B; FLJ35411; PP2A B subunit isoform B'-beta; PP2A B subunit isoform B56-beta; PP2A B subunit isoform PR61-beta; PP2A B subunit isoform R5-beta; PR61B; protein phosphatase 2 regulatory subunit B', beta; protein phosphatase 2, regulatory subunit B', beta isoform; serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, beta isoform; Serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit beta isoform
Gene Aliases: B56B; PPP2R5B; PR61B
UniProt ID: (Human) Q15173
Entrez Gene ID: (Human) 5526
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.