Invitrogen
Manufacturer Code:PA112387
Catalog # 01-674-120
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: PP2A B subunit isoform B'-alpha; PP2A B subunit isoform B56-alpha; PP2A B subunit isoform PR61-alpha; PP2A B subunit isoform R5-alpha; PP2A, B subunit, B' alpha isoform; PP2A, B subunit, B56 alpha isoform; PP2A, B subunit, PR61 alpha isoform; PP2A, B subunit, R5 alpha isoform; PP2CB; PR61alpha; protein phosphatase 2 regulatory subunit B', alpha; protein phosphatase 2, regulatory subunit B (B56), alpha isoform; protein phosphatase 2, regulatory subunit B', alpha isoform; serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, alpha isoform; Serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit alpha isoform
Gene Aliases: B56A; PPP2R5A; PR61A; PR61alpha
UniProt ID: (Human) Q15172, (Mouse) Q6PD03
Entrez Gene ID: (Human) 5525, (Mouse) 226849, (Rat) 312754
Molecular Function:
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.