Invitrogen
Manufacturer Code:PA5106900
Catalog # PIPA5106900
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PPP2R4 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610042B21Rik C77440 MGC2184 N28142 OTTHUMP00000022333 Phosphotyrosyl phosphatase activator PP2A PP2A phosphatase activator PP2A subunit B' isoform PR53 PP2A subunit B' PR53 isoform PPP2R4 PR53 protein phosphatase 2 phosphatase activator protein phosphatase 2 regulatory subunit 4 protein phosphatase 2A activator regulatory subunit 4 protein phosphatase 2A activator regulatory subunit B protein phosphatase 2A regulatory subunit 4 protein phosphatase 2A regulatory subunit B (PR 53) protein phosphatase 2A regulatory subunit B' (PR 53) PTPA Serine/threonine-protein phosphatase 2A activator serine/threonine-protein phosphatase 2A regulatory subunit 4 Serine/threonine-protein phosphatase 2A regulatory subunit B'; Phosphotyrosyl phosphatase activator; PP2A phosphatase activator; PP2A subunit B' isoform PR53; PP2A, subunit B', PR53 isoform; protein phosphatase 2 regulatory subunit 4; protein phosphatase 2A activator, regulatory subunit 4; protein phosphatase 2A regulatory subunit 4; protein phosphatase 2A, regulatory subunit B (PR 53); protein phosphatase 2A, regulatory subunit B' (PR 53); PTPA; Serine/threonine-protein phosphatase 2A activator; Serine/threonine-protein phosphatase 2A regulatory subunit 4; Serine/threonine-protein phosphatase 2A regulatory subunit B'
Gene Aliases: 2610042B21Rik; C77440; N28142; PP2A; PPP2R4; PR53; PTPA
UniProt ID: (Human) Q15257, (Mouse) P58389
Entrez Gene ID: (Human) 5524, (Rat) 362102, (Mouse) 110854
Molecular Function: enzyme modulator phosphatase activator phosphatase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.