Novus Biologicals
Manufacturer Code:NBP219909
Catalog # NBP219909
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human PPP2R4. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 5.2.1.8 MGC2184 Phosphotyrosyl phosphatase activator PP2A PP2A phosphatase activator PP2A subunit B' isoform PR53 PP2A subunit B' PR53 isoform protein phosphatase 2A activator regulatory subunit 4 regulatory subunit B' (PR 53) serine/threonine-protein phosphatase 2A activator Serine/threonine-protein phosphatase 2A regulatory subunit 4 Serine/threonine-protein phosphatase 2A regulatory subunit B'; Phosphotyrosyl phosphatase activator; PP2A phosphatase activator; PP2A subunit B' isoform PR53; PP2A, subunit B', PR53 isoform; protein phosphatase 2 regulatory subunit 4; protein phosphatase 2A activator, regulatory subunit 4; protein phosphatase 2A regulatory subunit 4; protein phosphatase 2A, regulatory subunit B' (PR 53); PTPA; Serine/threonine-protein phosphatase 2A activator; Serine/threonine-protein phosphatase 2A regulatory subunit 4; Serine/threonine-protein phosphatase 2A regulatory subunit B'
Gene Aliases: PP2A; PPP2R4; PR53; PTPA
UniProt ID: (Human) Q15257
Entrez Gene ID: (Human) 5524
Molecular Function:
enzyme modulator
phosphatase activator
phosphatase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.