R&D Systems
Manufacturer Code:AF5929SP
Catalog # AF5929SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <em>E. coli</em>-derived recombinant human PPP2R4 Glu14-Asn323 Accession No. Q15257 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Goat Polyclonal Antibody
Protein Aliases: EC 5.2.1.8 MGC2184 Phosphotyrosyl phosphatase activator PP2A PP2A phosphatase activator PP2A subunit B&prime isoform PR53 PP2A subunit B&prime PR53 isoform protein phosphatase 2A activator regulatory subunit 4 PTPA regulatory subunit B&prime (PR 53) serine/threonine-protein phosphatase 2A activator Serine/threonine-protein phosphatase 2A regulatory subunit 4 Serine/threonine-protein phosphatase 2A regulatory subunit B′ Phosphotyrosyl phosphatase activator; PP2A phosphatase activator; PP2A subunit B' isoform PR53; PP2A, subunit B', PR53 isoform; protein phosphatase 2 regulatory subunit 4; protein phosphatase 2A activator, regulatory subunit 4; protein phosphatase 2A regulatory subunit 4; protein phosphatase 2A, regulatory subunit B' (PR 53); PTPA; Serine/threonine-protein phosphatase 2A activator; Serine/threonine-protein phosphatase 2A regulatory subunit 4; Serine/threonine-protein phosphatase 2A regulatory subunit B'
Gene Aliases: PP2A; PPP2R4; PR53; PTPA
UniProt ID: (Human) Q15257
Entrez Gene ID: (Human) 5524
Molecular Function: enzyme modulator phosphatase activator phosphatase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.