Proteintech
Manufacturer Code:248771AP150UL
Catalog # 50-173-4591
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PPP2R3C Fusion Protein Ag19282 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C14orf10 G4 1 G5pr PPP2R3C; phosphatase subunit gene g4-1; protein phosphatase 2 (formerly 2A), regulatory subunit B''; protein phosphatase 2 regulatory subunit B'', gamma; protein phosphatase 2, regulatory subunit B'', gamma; Protein phosphatase subunit G5PR; rhabdomyosarcoma antigen MU-RMS-40.6A; Rhabdomyosarcoma antigen MU-RMS-40.6A/6C; rhabdomyosarcoma antigen Mu-RMS-40.6C; Serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit gamma
Gene Aliases: 4930511A21Rik; 5730412A08Rik; C14orf10; G4-1; G5PR; MNCb-1932; PPP2R3C; RGD1309207
UniProt ID: (Human) Q969Q6, (Mouse) Q9JK24, (Rat) Q6AXZ3
Entrez Gene ID: (Human) 55012, (Mouse) 59032, (Rat) 362739
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.